Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(n-Nonyloxy)benzoic acid, 97%, Thermo Scientific™

Catalog No. AAB2049606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2049606 5 g
AAB2049603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2049606 Supplier Thermo Scientific Chemicals Supplier No. B2049606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15872-43-2
Molecular Formula C16H24O3
Molecular Weight (g/mol) 264.365
MDL Number MFCD00043501
InChI Key BOZLUAUKDKKZHJ-UHFFFAOYSA-N
Synonym p-nonyloxybenzoic acid, 4-nonyloxy benzoic acid, 4-n-nonyloxybenzoic acid, 4-n-nonyloxy benzoic acid, 4-nonyloxybenzoic acid, 4-nonyloxy benzoic acid #, benzoic acid,4-nonyloxy, rarechem al be 1496, timtec-bb sbb008264
PubChem CID 139992
IUPAC Name 4-nonoxybenzoic acid
SMILES CCCCCCCCCOC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point ∼143°C
Quantity 5 g
Beilstein 2213560
Formula Weight 264.37
Percent Purity 97%
Chemical Name or Material 4-(n-Nonyloxy)benzoic acid
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.