CAS RN 15872-44-3
IUPAC Name:
4-(undecyloxy)benzoic acid
Synonyms:
4-(undecyloxy)benzoic acid
Molecular Weight (g/mol):
292.42
Molecular Formula:
C18H28O3
InChi Key:
NEJZHJHZOUISSH-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCOC1=CC=C(C=C1)C(O)=O
1
–
1
of
1
results
1
4-Undecyloxybenzoic Acid 98.0+%, TCI America™
CAS: 15872-44-3 Molecular Formula: C18H28O3 Molecular Weight (g/mol): 292.419 MDL Number: MFCD00043772 InChI Key: NEJZHJHZOUISSH-UHFFFAOYSA-N Synonym: 4-undecyloxy benzoic acid,4-undecyloxybenzoic acid,4-n-undecyloxybenzoic acid,p-undecyloxybenzoic acid,pubchem3929,4-undecyloxybenzoicacid,acmc-209diw,4-undecyloxy benzoic acid #,benzoic acid,4-undecyloxy,timtec-bb sbb008333 PubChem CID: 519187 IUPAC Name: 4-undecoxybenzoic acid SMILES: CCCCCCCCCCCOC1=CC=C(C=C1)C(=O)O