Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Undecyloxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. U00725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00725G Supplier TCI America Supplier No. U00725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15872-44-3
Molecular Formula C18H28O3
Molecular Weight (g/mol) 292.419
MDL Number MFCD00043772
InChI Key NEJZHJHZOUISSH-UHFFFAOYSA-N
Synonym 4-undecyloxy benzoic acid, 4-undecyloxybenzoic acid, 4-n-undecyloxybenzoic acid, p-undecyloxybenzoic acid, pubchem3929, 4-undecyloxybenzoicacid, acmc-209diw, 4-undecyloxy benzoic acid #, benzoic acid,4-undecyloxy, timtec-bb sbb008333
PubChem CID 519187
IUPAC Name 4-undecoxybenzoic acid
SMILES CCCCCCCCCCCOC1=CC=C(C=C1)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 292.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Undecyloxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.