CAS RN 15873-72-0
IUPAC Name:
dicyclohexylphosphinoyl chloride
Synonyms:
dicyclohexylphosphinoyl chloride
Molecular Weight (g/mol):
248.73
Molecular Formula:
C12H22ClOP
InChi Key:
HKYSIUYOULVHTO-UHFFFAOYSA-N
SMILES:
ClP(=O)(C1CCCCC1)C1CCCCC1
1
–
1
of
1
results
1
Dicyclohexylphosphinyl Chloride 98.0+%, TCI America™
CAS: 15873-72-0 Molecular Formula: C12H22ClOP Molecular Weight (g/mol): 248.731 MDL Number: MFCD00191344 InChI Key: HKYSIUYOULVHTO-UHFFFAOYSA-N PubChem CID: 557982 IUPAC Name: [chloro(cyclohexyl)phosphoryl]cyclohexane SMILES: C1CCC(CC1)P(=O)(C2CCCCC2)Cl