accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 1588-89-2

IUPAC Name: 2,3-diphenylpyrazine
Synonyms: 2,3-diphenylpyrazine
Molecular Weight (g/mol): 232.29
Molecular Formula: C16H12N2
InChi Key: PTZIVVDMBCVSMR-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=NC=CN=C1C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)