accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OH

CAS RN 15890-36-5

IUPAC Name: 4-(propan-2-yl)cyclohexan-1-ol
Synonyms: isopropylcyclohexanol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: DKKRDMLKVSKFMJ-UHFFFAOYSA-N
SMILES: CC(C)C1CCC(O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results