Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Isopropylcyclohexanol 93.0+%, TCI America™
SDP

Catalog No. I07821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07821G Supplier TCI America Supplier No. I07821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15890-36-5
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
InChI Key DKKRDMLKVSKFMJ-UHFFFAOYSA-N
PubChem CID 20739
IUPAC Name 4-propan-2-ylcyclohexan-1-ol
SMILES CC(C)C1CCC(CC1)O

Specifications

Melting Point 6°C
Color Colorless
Boiling Point 94°C
Quantity 1 g
Formula Weight 142.24
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-4-Isopropylcyclohexanol
EINECSNumber (3)-2301
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.