CAS RN 15890-72-9
CAS RN 15890-72-9
IUPAC Name:
magnesium(2+) dodecan-1-ide bromide
Synonyms:
magnesium(2+) dodecan-1-ide bromide
Molecular Weight (g/mol):
273.54
Molecular Formula:
C12H25BrMg
InChi Key:
MBHQEXPGNCWWBP-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].CCCCCCCCCCC[CH2-]
1
–
1
of
1
results
1
Sigma Aldrich Dodecylmagnesium Bromide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.