accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 158937-25-8

IUPAC Name: [4'-(pentyloxy)-[1,1'-biphenyl]-4-yl]boronic acid
Synonyms: 4'-(pentyloxy)-[1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol): 284.16
Molecular Formula: C17H21BO3
InChi Key: DBDYXLZXTNHAFI-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)B(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results