Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Pentyloxybiphenyl-4-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P23655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2365-5G 5 g
P23651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2365-5G Supplier TCI America Supplier No. P23655G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 158937-25-8
Molecular Formula C17H21BO3
Molecular Weight (g/mol) 284.16
MDL Number MFCD07644475
InChI Key DBDYXLZXTNHAFI-UHFFFAOYSA-N
PubChem CID 11358071
IUPAC Name [4'-(pentyloxy)-[1,1'-biphenyl]-4-yl]boronic acid
SMILES CCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 197°C
Color White
Quantity 5 g
Synonym 4 'C-Amyloxybiphenyl-4-boronic Acid, [4 'C-(Pentyloxy)-[1,1 'C-biphenyl]-4-yl]boronic Acid
Formula Weight 284.16
Physical Form Crystalline Powder
Chemical Name or Material 4 '-Pentyloxybiphenyl-4-boronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.