accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSBrBr

CAS RN 158956-23-1

IUPAC Name: 2,5-dibromo-3-decylthiophene
Synonyms: 2,5-dibromo-3-decylthiophene
Molecular Weight (g/mol): 382.2
Molecular Formula: C14H22Br2S
InChi Key: CQDSRZZKSUNTSB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=C(Br)SC(Br)=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)