Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-3-decylthiophene 97.0+%, TCI America™
SDP

Catalog No. D45725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45725G 5 g
D457225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45725G Supplier TCI America Supplier No. D45725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 158956-23-1
Molecular Formula C14H22Br2S
Molecular Weight (g/mol) 382.20
MDL Number MFCD00274314
InChI Key CQDSRZZKSUNTSB-UHFFFAOYSA-N
Synonym 2,5-dibromo-3-decyl-thiophene, pubchem22579, bidd:gt0393, 2,5-dibromo-3-n-decylthiophene, thiophene, 2,5-dibromo-3-decyl, 2,5-bis bromanyl-3-decyl-thiophene, 2,5-dibromo-3-decylthiophene
PubChem CID 4218617
IUPAC Name 2,5-dibromo-3-decylthiophene
SMILES CCCCCCCCCCC1=C(Br)SC(Br)=C1

Specifications

Boiling Point 148°C
Quantity 5 g
Formula Weight 382.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dibromo-3-decylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.