CAS RN 159002-17-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
N(alpha)-Boc-DL-2,3-diaminopropionic acid, 97%
N(alpha)-Boc-DL-2,3-diaminopropionic acid, 97%
CAS: 159002-17-2 Molecular Formula: C8H16N2O4 MDL Number: MFCD00736752 Synonym: Boc-DL-Dap-OH; 3-Amino-N-Boc-DL-alanine