accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15908-64-2

IUPAC Name: 4-(methylsulfanyl)-6-oxo-2-phenyl-1,6-dihydropyrimidine-5-carbonitrile
Synonyms: 4-(methylsulfanyl)-6-oxo-2-phenyl-1H-pyrimidine-5-carbonitrile
Molecular Weight (g/mol): 243.28
Molecular Formula: C12H9N3OS
InChi Key: OUSBLYHTCSUCNF-UHFFFAOYSA-N
SMILES: CSC1=C(C#N)C(=O)NC(=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results