accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1592-38-7

IUPAC Name: (naphthalen-2-yl)methanol
Synonyms: 2-naphthalenemethanol
Molecular Weight (g/mol): 158.2
Molecular Formula: C11H10O
InChi Key: MFGWMAAZYZSWMY-UHFFFAOYSA-N
SMILES: OCC1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)