Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthalenemethanol 98.0+%, TCI America™
SDP

Catalog No. N08815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08815G 5 g
N088125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08815G Supplier TCI America Supplier No. N08815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1592-38-7
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004124
InChI Key MFGWMAAZYZSWMY-UHFFFAOYSA-N
Synonym 2-naphthalenemethanol, 2-naphthylmethanol, 2-hydroxymethylnaphthalene, 2-naphthalene methanol, 2-naphthyl methanol, naphthalen-2-yl-methanol, naphthalen-2-yl methanol, 2-naphthylmethan-1-ol, 2-naphthylcarbinol, 2na
PubChem CID 74128
ChEBI CHEBI:27615
IUPAC Name naphthalen-2-ylmethanol
SMILES C1=CC=C2C=C(C=CC2=C1)CO

Specifications

Melting Point 80°C
Color White-Yellow
Boiling Point 178°C
Quantity 5 g
Formula Weight 158.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthalenemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.