CAS RN 159326-68-8
IUPAC Name:
pyrrolo[2,1-f][1,2,4]triazin-4-amine
Synonyms:
pyrrolo[2,1-f][1,2,4]triazin-4-amine
Molecular Weight (g/mol):
134.14
Molecular Formula:
C6H6N4
InChi Key:
VSPXQZSDPSOPRO-UHFFFAOYSA-N
SMILES:
NC1=NC=NN2C=CC=C12
1
–
1
of
1
results
1
Promotions Available
Pyrrolo[2,1-f][1,2,4]triazin-4-amine, Thermo Scientific Chemicals
Pyrrolo[2,1-f][1,2,4]triazin-4-amine, Thermo Scientific Chemicals
CAS: 159326-68-8 Molecular Formula: C6H6N4 Molecular Weight (g/mol): 134.14 MDL Number: MFCD08234647 InChI Key: VSPXQZSDPSOPRO-UHFFFAOYSA-N IUPAC Name: pyrrolo[2,1-f][1,2,4]triazin-4-amine SMILES: NC1=NC=NN2C=CC=C12