Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrolo[2,1-f][1,2,4]triazin-4-amine, Thermo Scientific Chemicals

Catalog No. AC468990050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468990050 5 g
AC468990010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468990050 Supplier Thermo Scientific Chemicals Supplier No. 468990050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 159326-68-8 | C6H6N4 | 134.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159326-68-8
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.14
MDL Number MFCD08234647
InChI Key VSPXQZSDPSOPRO-UHFFFAOYSA-N
IUPAC Name pyrrolo[2,1-f][1,2,4]triazin-4-amine
SMILES NC1=NC=NN2C=CC=C12

Specifications

Quantity 5 g
Formula Weight 134.12
Chemical Name or Material Pyrrolo[2,1-f][1,2,4 ]triazin-4-amine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May cause an allergic skin reaction.

Causes serious eye irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.