accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1594-57-6

IUPAC Name: (Z)-N'-hydroxypyridine-4-carboximidamide
Synonyms: (Z)-N'-hydroxypyridine-4-carboximidamide
Molecular Weight (g/mol): 137.14
Molecular Formula: C6H7N3O
InChi Key: ZUUATXHRJFHSGO-UHFFFAOYSA-N
SMILES: N\C(=N/O)C1=CC=NC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)