Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N'-Hydroxypyridine-4-carboximidamide, 97%, Thermo Scientific™

Catalog No. AC432480050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432480050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC432480050 Supplier Thermo Scientific Chemicals Supplier No. 432480050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1594-57-6
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00125873
InChI Key ZUUATXHRJFHSGO-UHFFFAOYSA-N
Synonym n'-hydroxypyridine-4-carboximidamide, pyridine-4-amidoxime, 4-pyridylamidoxime, n-hydroxyisonicotinamidine, n-hydroxy-isonicotinamidine, isonicotinamidoxime, isonicotinohydroxamamide, 4-pyridinecarboxamidoxime, n-hydroxyisonicotinimidamide, z-n'-hydroxyisonicotinimidamide
PubChem CID 5355713
IUPAC Name nitroso(1H-pyridin-4-ylidene)methanamine
SMILES N\C(=N/O)C1=CC=NC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 175.0°C to 180.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 137.14
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material N'-Hydroxypyridine-4-carboximidamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.