accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFH2N

CAS RN 1597-33-7

IUPAC Name: 2-fluoropyridin-3-amine
Synonyms: 2-fluoropyridin-3-amine
Molecular Weight (g/mol): 112.11
Molecular Formula: C5H5FN2
InChi Key: QVDACIZNNNFTBO-UHFFFAOYSA-N
SMILES: NC1=C(F)N=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results