Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2-fluoropyridine 98.0+%, TCI America™
SDP

Catalog No. A26501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26501G Supplier TCI America Supplier No. A26501G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1597-33-7
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.107
MDL Number MFCD03095248
InChI Key QVDACIZNNNFTBO-UHFFFAOYSA-N
PubChem CID 2762816
IUPAC Name 2-fluoropyridin-3-amine
SMILES C1=CC(=C(N=C1)F)N

Specifications

Boiling Point 102°C
Quantity 1 g
Formula Weight 112.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Amino-2-fluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.