accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15971-92-3

IUPAC Name: ethyl 3-cyclohexyl-3-oxopropanoate
Synonyms: ethyl 3-cyclohexyl-3-oxopropanoate
Molecular Weight (g/mol): 198.26
Molecular Formula: C11H18O3
InChi Key: ASYASKBLHYSMEG-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results