Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Cyclohexyl-3-oxopropionate 98.0+%, TCI America™
SDP

Catalog No. E11261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E11261G Supplier TCI America Supplier No. E11261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15971-92-3
Molecular Formula C11H18O3
Molecular Weight (g/mol) 198.262
MDL Number MFCD04115574
InChI Key ASYASKBLHYSMEG-UHFFFAOYSA-N
Synonym 3-Cyclohexyl-3-oxopropionic Acid Ethyl Ester
PubChem CID 266327
IUPAC Name ethyl 3-cyclohexyl-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1CCCCC1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 1 g
Formula Weight 198.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Cyclohexyl-3-oxopropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.