accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH3C

CAS RN 15987-00-5

IUPAC Name: 2-butyl-3-methylpyrazine
Synonyms: pyrazine, 2-butyl-3-methyl-
Molecular Weight (g/mol): 150.23
Molecular Formula: C9H14N2
InChi Key: IRSIKJPEFMMRHD-UHFFFAOYSA-N
SMILES: CCCCC1=NC=CN=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results