Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butyl-3-methylpyrazine 98.0+%, TCI America™
SDP

Catalog No. B11731ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11731ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11731ML Supplier TCI America Supplier No. B11731ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15987-00-5
Molecular Formula C9H14N2
Molecular Weight (g/mol) 150.23
MDL Number MFCD00038031
InChI Key IRSIKJPEFMMRHD-UHFFFAOYSA-N
PubChem CID 85228
IUPAC Name 2-butyl-3-methylpyrazine
SMILES CCCCC1=NC=CN=C1C

Specifications

Boiling Point 84°C
Quantity 1 mL
Formula Weight 150.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Butyl-3-methylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.