CAS RN 160032-40-6
IUPAC Name:
{thieno[3,2-b]thiophen-2-yl}boronic acid
Synonyms:
thieno[3,2-b]thiophen-2-ylboronic acid
Molecular Weight (g/mol):
184.03
Molecular Formula:
C6H5BO2S2
InChi Key:
PVQAASIVXYZXFF-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC2=C(S1)C=CS2
1
–
1
of
1
results
1
Thieno[3,2-b]thiophene-2-boronic Acid (contains varying amounts of Anhydride), TCI America™
CAS: 160032-40-6 Molecular Formula: C6H5BO2S2 Molecular Weight (g/mol): 184.034 InChI Key: PVQAASIVXYZXFF-UHFFFAOYSA-N PubChem CID: 10035286 IUPAC Name: thieno[3,2-b]thiophen-5-ylboronic acid SMILES: B(C1=CC2=C(S1)C=CS2)(O)O