Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thieno[3,2-b]thiophene-2-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. T26211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26211G Supplier TCI America Supplier No. T26211G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 160032-40-6
Molecular Formula C6H5BO2S2
Molecular Weight (g/mol) 184.034
InChI Key PVQAASIVXYZXFF-UHFFFAOYSA-N
PubChem CID 10035286
IUPAC Name thieno[3,2-b]thiophen-5-ylboronic acid
SMILES B(C1=CC2=C(S1)C=CS2)(O)O

Specifications

Quantity 1 g
Formula Weight 184.03
Physical Form Crystalline Powder
Chemical Name or Material Thieno[3,2-b]thiophene-2-boronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.