accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNCH3CH2CH3H2CH3CHOOHOOFeCl(Z)(Z)(Z)(Z)

CAS RN 16009-13-5

IUPAC Name: 3-[(1Z,6Z,12Z,17Z)-5-(2-carboxyethyl)-22-chloro-15,20-diethenyl-4,10,14,19-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,12,14,16(24),17,19-undecaen-9-yl]propanoic acid
Synonyms: 3-[(1Z,6Z,12Z,17Z)-5-(2-carboxyethyl)-22-chloro-15,20-diethenyl-4,10,14,19-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,12,14,16(24),17,19-undecaen-9-yl]propanoic acid
Molecular Weight (g/mol): 651.95
Molecular Formula: C34H32ClFeN4O4
InChi Key: BTIJJDXEELBZFS-HXFTUNQESA-K
SMILES: CC1=C(CCC(O)=O)\C2=C\C3=C(CCC(O)=O)C(C)=C4\C=C5/N=C(/C=C6\N([Fe](Cl)N34)\C(=C/C1=N2)C(C=C)=C6C)C(C=C)=C5C
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)