Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemin Chloride, MP Biomedicals™

Catalog No. MP21988202 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21988202 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21988202 Supplier MP Biomedicals, Inc Supplier No. 02198820.2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to study bacterial strains and growth conditions. Hemin Chloride, MP Biomedicals is the oxidized version of heme. It is an iron-containing porphyrin, a prosthetic group for a diverse group of proteins. Free heme is pro-inflammatory and contributes to

Hemin is a Protoporphyrin IX containing a ferric iron ion (Heme B) with a chloride ligand. It is the oxidized version of heme. Free heme can be released from hemoglobin following hemolysis. Free hemin levels may be upregulated in various pathological conditions and can contribute to various inflammatory conditions including vascular disorders, renal failure, and immune-mediated disorders.

Hemin is used for studying bacterial strains and growth conditions, cell transfections, and short-term colony assays (the culture medium consists of hemin along with other components). It is used in β-hematin formation assay for analyzing the inhibitory activity of MMV (Medicines for Malaria Venture) Malaria Box compounds against the formation of β-hematin. It may also be used to study the expression, represson and activity of heme oxygenase 1 (HO-1). Hematin is the “X factor” required to grown Haemophilus influenzae.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16009-13-5
Molecular Formula C34H32ClFeN4O4
Molecular Weight (g/mol) 651.95
MDL Number MFCD00010726
InChI Key BTIJJDXEELBZFS-HXFTUNQESA-K
Synonym chlorohemin, hemin chloride, protoferriheme, ferriheme, ferriprotoporphyrin ix chloride, ferriprotoporphyrin ix, panhematin, protohemin, protohemin ix, ferriprotoporphyrin
PubChem CID 131675604
IUPAC Name 3-[(1Z,6Z,12Z,17Z)-5-(2-carboxyethyl)-22-chloro-15,20-diethenyl-4,10,14,19-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,12,14,16(24),17,19-undecaen-9-yl]propanoic acid
SMILES CC1=C(CCC(O)=O)\C2=C\C3=C(CCC(O)=O)C(C)=C4\C=C5/N=C(/C=C6\N([Fe](Cl)N34)\C(=C/C1=N2)C(C=C)=C6C)C(C=C)=C5C

Specifications

Melting Point 2735°C (1013hPa (literature))
Quantity 250 mg
Formula Weight 652g/mol
Physical Form Powder
Recommended Storage Store at 4°C, protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.