accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 160168-40-1

CAS RN 160168-40-1

IUPAC Name: (2S,3R)-3-{[(2R,3R,4R,5S,6R)-4,5-bis(acetyloxy)-6-[(acetyloxy)methyl]-3-acetamidooxan-2-yl]oxy}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: (2S,3R)-3-{[(2R,3R,4R,5S,6R)-4,5-bis(acetyloxy)-6-[(acetyloxy)methyl]-3-acetamidooxan-2-yl]oxy}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 670.67
Molecular Formula: C33H38N2O13
InChi Key: OXLCJWGAUPPZQJ-WPYJLJGXSA-N
SMILES: C[C@@H](O[C@@H]1O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]1NC(C)=O)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)