accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOHO

CAS RN 16024-58-1

IUPAC Name: 2-[2-(2-methoxyethoxy)ethoxy]acetic acid
Synonyms: [2-(2-methoxyethoxy)ethoxy]acetic acid
Molecular Weight (g/mol): 178.18
Molecular Formula: C7H14O5
InChi Key: YHBWXWLDOKIVCJ-UHFFFAOYSA-N
SMILES: COCCOCCOCC(O)=O
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)