accessibility menu, dialog, popup

UserName

Viewing profile as user:
OCrOOClK

CAS RN 16037-50-6

IUPAC Name: potassium trioxochromium chloride
Synonyms: potassium chromium trioxide chloride
Molecular Weight (g/mol): 174.54
Molecular Formula: ClCrKO3
InChi Key: PEBPTQFCMJWPGZ-UHFFFAOYSA-M
SMILES: [Cl-].[K+].O=[Cr](=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results