CAS RN 16037-91-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Sodium Stibogluconate
Sodium Stibogluconate, CAS 16037-91-5, is a pentavalent antimony compound that irreversibly inhibits protein tyrosine phosphatase activity, including SHP-1, by forming a stable complex.