Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Sodium Stibogluconate

Catalog No. 5675651GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-756-51GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-756-51GM Supplier MilliporeSigma™ Supplier No. 5675651GM
Add to Cart
Add to Cart

Sodium Stibogluconate, CAS 16037-91-5, is a pentavalent antimony compound that irreversibly inhibits protein tyrosine phosphatase activity, including SHP-1, by forming a stable complex.

A pentavalent antimony compound that irreversibly inhibits protein tyrosine phosphatase (PTPase) activity, including Src homology PTPase-1 (SHP-1) (99% inhibition at ∽11μM) by forming a stable complex. At higher concentrations (∽110μM), it inhibits SHP-2 and PTP1B activities.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 90% by titration on dry basis (30% Sb content)
Target Proteins PTPase activity, including Src homology PTPase-1 (SHP-1)
CAS 16037-91-5
Molecular Formula C12H17O17Sb2·3Na·9H2O
Formula Weight 907.9g/mol
Solubility H2O
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.