accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONN

CAS RN 160446-18-4

IUPAC Name: methyl 4-[(1H-imidazol-1-yl)methyl]benzoate
Synonyms: methyl 4-(imidazol-1-ylmethyl)benzoate
Molecular Weight (g/mol): 216.24
Molecular Formula: C12H12N2O2
InChi Key: HMWIIHIOSBESMH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(CN2C=CN=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results