Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-(1H-imidazol-1-ylmethyl)benzoate, 97%, Thermo Scientific™

Catalog No. CC43322DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC43322DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC43322DA Supplier Thermo Scientific Chemicals Supplier No. CC43322DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 160446-18-4
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD00608318
InChI Key HMWIIHIOSBESMH-UHFFFAOYSA-N
Synonym methyl 4-1h-imidazol-1-ylmethyl benzoate, methyl 4-imidazol-1-ylmethyl benzoate, methyl 4-1h-imidazol-1-yl methyl benzoate, methyl 4-imidazolylmethyl benzoate, 4-imidazol-1-ylmethyl-benzoic acid methyl ester, 4-1h-imidazol-1-ylmethyl benzoic acid methyl ester, benzoicacid, 4-1h-imidazol-1-ylmethyl-, methyl ester
PubChem CID 3164140
IUPAC Name methyl 4-(imidazol-1-ylmethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)CN2C=CN=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material Methyl 4-(1H-imidazol-1-ylmethyl)benzoate
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.