CAS RN 160450-10-2
CAS RN 160450-10-2
IUPAC Name:
methyl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1H-imidazol-5-yl)propanoate
Synonyms:
methyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(3H-imidazol-4-yl)propanoate
Molecular Weight (g/mol):
391.43
Molecular Formula:
C22H21N3O4
InChi Key:
YJYNUBXPJQOMJC-FQEVSTJZSA-N
SMILES:
COC(=O)[C@H](CC1=CN=CN1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-His-OMe, Advanced ChemTech
CAS: 160450-10-2 Molecular Formula: C22H21N3O4 Molecular Weight (g/mol): 391.43 MDL Number: MFCD27578253 InChI Key: YJYNUBXPJQOMJC-FQEVSTJZSA-N IUPAC Name: methyl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1H-imidazol-5-yl)propanoate SMILES: COC(=O)[C@H](CC1=CN=CN1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2