accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)(R)CH3(R)Cl

CAS RN 16052-42-9

IUPAC Name: (1S,2R,4R)-2-chloro-4-methyl-1-(propan-2-yl)cyclohexane
Synonyms: (1S,2R,4R)-2-chloro-1-isopropyl-4-methylcyclohexane
Molecular Weight (g/mol): 174.71
Molecular Formula: C10H19Cl
InChi Key: OMLOJNNKKPNVKN-KXUCPTDWSA-N
SMILES: CC(C)[C@@H]1CC[C@@H](C)C[C@H]1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results