Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Menthyl Chloride 98.0+%, TCI America™
SDP

Catalog No. M08341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M08341G Supplier TCI America Supplier No. M08341G

Chemical Identifiers

CAS 16052-42-9
Molecular Formula C10H19Cl
Molecular Weight (g/mol) 174.712
MDL Number MFCD00001481
InChI Key OMLOJNNKKPNVKN-KXUCPTDWSA-N
Synonym (-)-3-Chloro-p-menthane
PubChem CID 244509
IUPAC Name (1S,2R,4R)-2-chloro-4-methyl-1-propan-2-ylcyclohexane
SMILES CC1CCC(C(C1)Cl)C(C)C

Specifications

Color Colorless
Boiling Point 101°C
Quantity 1 g
Formula Weight 174.71
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-Menthyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.