accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNClCl

CAS RN 16063-70-0

IUPAC Name: 2,3,5-trichloropyridine
Synonyms: 2,3,5-trichloropyridine
Molecular Weight (g/mol): 182.43
Molecular Formula: C5H2Cl3N
InChi Key: CNLIIAKAAMFCJG-UHFFFAOYSA-N
SMILES: ClC1=CN=C(Cl)C(Cl)=C1
Boiling Point 
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)