Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Trichloropyridine 98.0+%, TCI America™
SDP

Catalog No. T147925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T147925G 25 g
T1479250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T147925G Supplier TCI America Supplier No. T147925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16063-70-0 | C5H2Cl3N | 182.428 g/mol

Chemical Identifiers

CAS 16063-70-0
Molecular Formula C5H2Cl3N
Molecular Weight (g/mol) 182.428
MDL Number MFCD00043007
InChI Key CNLIIAKAAMFCJG-UHFFFAOYSA-N
Synonym pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
PubChem CID 27666
IUPAC Name 2,3,5-trichloropyridine
SMILES C1=C(C=NC(=C1Cl)Cl)Cl

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 220°C
Quantity 25 g
Formula Weight 182.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5-Trichloropyridine
EINECSNumber (5)-5258
RTECSNumber UU0525000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.