accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHSOHSH

CAS RN 16096-97-2

IUPAC Name: 1,4-disulfanylbutane-2,3-diol
Synonyms: dithiotreitol DTE dithioerythreitol cleland's reagent rac-dithiothreitol
Molecular Weight (g/mol): 154.24
Molecular Formula: C4H10O2S2
InChi Key: VHJLVAABSRFDPM-UHFFFAOYNA-N
SMILES: OC(CS)C(O)CS
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)