Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dithio-L-threitol 99%, Thermo Scientific™

Catalog No. AC204365000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC204365000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC204365000 Supplier Thermo Scientific Chemicals Supplier No. 204365000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16096-97-2
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00064305
InChI Key VHJLVAABSRFDPM-UHFFFAOYNA-N
Synonym l-dithiothreitol, l-dtt, dithiothreitol, 1,4-dithiothreitol, l-1,4-dithiothreitol, cleland reagent chiral, l---dithiothreitol, 2,3-dihydroxy-1,4-dithiobutane, 2r,3r-1,4-dimercapto-2,3-butanediol, rel-2r,3r-1,4-dimercaptobutane-2,3-diol
PubChem CID 439196
ChEBI CHEBI:42106
IUPAC Name 1,4-disulfanylbutane-2,3-diol
SMILES OC(CS)C(O)CS

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50°C to 54°C
Color White
Infrared Spectrum Authentic
Linear Formula HSCH2CH(OH)CH(OH)CH2SH
Quantity 500 mg
Merck Index 10, 3394
Specific Rotation -13.5° to -16.5° (20°C, 589nm) (c=2, CHCl3)
Formula Weight 154.24
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 1, 4-Dithio-L-threitol, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.