accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOOOFFF

CAS RN 160969-03-9

IUPAC Name: 2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl methanesulfonate
Synonyms: 2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl methanesulfonate
Molecular Weight (g/mol): 314.28
Molecular Formula: C11H13F3O5S
InChi Key: HOJMCBMXHWZNKX-UHFFFAOYSA-N
SMILES: CS(=O)(=O)OCCOC1=CC=CC=C1OCC(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results