accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNHCH3CH3NNHH3CCH3N

CAS RN 1610-18-0

IUPAC Name: 6-methoxy-N2,N4-bis(propan-2-yl)-1,3,5-triazine-2,4-diamine
Synonyms: ontrack methoxypropazine prometone pramitol gesafram prometon
Molecular Weight (g/mol): 225.3
Molecular Formula: C10H19N5O
InChi Key: ISEUFVQQFVOBCY-UHFFFAOYSA-N
SMILES: COC1=NC(NC(C)C)=NC(NC(C)C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results