accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16111-07-2

IUPAC Name: 2-{3',3'-dimethyl-6-nitro-1',3'-dihydrospiro[chromene-2,2'-indol]-1'-yl}ethan-1-ol
Synonyms: 2-{3',3'-dimethyl-6-nitrospiro[chromene-2,2'-indol]-1'-yl}ethanol
Molecular Weight (g/mol): 352.39
Molecular Formula: C20H20N2O4
InChi Key: FGJPKPQMQFZPJY-UHFFFAOYNA-N
SMILES: CC1(C)C2=CC=CC=C2N(CCO)C11OC2=C(C=C1)C=C(C=C2)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results