Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Hydroxyethyl)-3,3-dimethylindolino-6'-nitrobenzopyrylospiran 93.0+%, TCI America™
SDP

Catalog No. H10425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10425G 5 g
H10421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10425G Supplier TCI America Supplier No. H10425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16111-07-2
Molecular Formula C20H20N2O4
Molecular Weight (g/mol) 352.39
MDL Number MFCD00369031
InChI Key FGJPKPQMQFZPJY-UHFFFAOYSA-N
PubChem CID 3104858
IUPAC Name 2-(3',3'-dimethyl-6-nitrospiro[chromene-2,2'-indole]-1'-yl)ethanol
SMILES CC1(C2=CC=CC=C2N(C13C=CC4=C(O3)C=CC(=C4)[N+](=O)[O-])CCO)C

Specifications

Melting Point 165°C
Color Purple-Red
Quantity 5 g
Synonym 1′-(2-Hydroxyethyl)-3′,3′-dimethyl-6-nitrospiro[1(2H)-benzopyran-2,2′-indoline]
Formula Weight 352.39
Percent Purity ≥93.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Hydroxyethyl)-3,3-dimethylindolino-6′-nitrobenzopyrylospiran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.