accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHO(S)CH3OOHN

CAS RN 16118-49-3

IUPAC Name: (1S)-1-(ethylcarbamoyl)ethyl N-phenylcarbamate
Synonyms: (1S)-1-(ethylcarbamoyl)ethyl N-phenylcarbamate
Molecular Weight (g/mol): 236.27
Molecular Formula: C12H16N2O3
InChi Key: AMRQXHFXNZFDCH-VIFPVBQESA-N
SMILES: CCNC(=O)[C@H](C)OC(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results