accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1612793-07-3

IUPAC Name: 2-[10-(dicyanomethylidene)-3-ethenyl-9,10-dihydroanthracen-9-ylidene]propanedinitrile
Synonyms: 2-[10-(dicyanomethylidene)-3-ethenylanthracen-9-ylidene]propanedinitrile
Molecular Weight (g/mol): 330.35
Molecular Formula: C22H10N4
InChi Key: SUTLXPZYDBOIRV-UHFFFAOYSA-N
SMILES: C=CC1=CC=C2C(=C1)C(=C(C#N)C#N)C1=CC=CC=C1C2=C(C#N)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results